Essigsäureanhydrid-[13C4,D6] (99% ¹³C , 97% D), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acetic anhydride-[¹³C₄,²H₆]

M-Clarity™ Qualitätsstufe = MQ200

607428-100MG
SIAL607428-100MGEA 1110 EUR
SIAL607428-100MG
Essigsäureanhydrid-[13C4,D6] (99% ¹³C , 97% D), Sigma-Aldrich®
Essigsäureanhydrid-[13C4,D6]
Formel: C₄H₆O₃
MDL: MFCD00190254
CAS-Nummer: 285977-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR