Essigsäureanhydrid-[1,1'-13C2,D6] (99% ¹³C , 98% D), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Acetic anhydride-[1,1'-¹³C₂,²H₆]

M-Clarity™ Qualitätsstufe = MQ200

607452-100MG
SIAL607452-100MGEA 986 EUR
SIAL607452-100MG
Essigsäureanhydrid-[1,1'-13C2,D6] (99% ¹³C , 98% D), Sigma-Aldrich®
Essigsäureanhydrid-[1,1'-13C2,D6]
Formel: C₄H₆O₃
MDL: MFCD00190252
CAS-Nummer: 285977-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR