L(+)-Methyl-tert-leucinat Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl

61891-1G
SIAL61891-1GEA 328 EUR
SIAL61891-1G
L(+)-Methyl-tert-leucinat Hydrochlorid, Sigma-Aldrich®
L(+)-Methyl-tert-leucinat Hydrochlorid
Formel: C₇H₁₅NO₂·HCl
Molecular Weight: 181,7 g/mol
MDL: MFCD00040604
CAS-Nummer: 63038-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR