(R)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

61902-500MG-F
SIAL61902-500MG-FEA 236 EUR
SIAL61902-500MG-F
(R)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(4-chlorphenyl)propionsäure
Formel: C₁₄H₁₈ClNO₄
Molecular Weight: 299,7500 g/mol
MDL: MFCD03427960
CAS-Nummer: 479064-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR