Lithiummethanolat 10% in Methanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Lithiummethylat, Methanol Lithiumsalz, Lithiummethoxid
62562-100ML-F
SIAL62562-100ML-FEA
240
EUR
SIAL62562-100ML-F
Lithiummethanolat 10% in Methanol, Sigma-Aldrich®
Lithiummethanolat
Formel:
CH₃LiO Molecular Weight: 37,98 g/mol |
MDL:
MFCD00036357 CAS-Nummer: 865-34-9 EINECS: 212-737-7 UN: 3274 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...