6-Hydroxy-3-coumaranon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Hydroxybenzofuran-3(2H)-one, 6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one

M-Clarity™ Qualitätsstufe = MQ100

630039-5G
SIAL630039-5GEA 34 EUR
SIAL630039-5G
6-Hydroxy-3-coumaranon, Sigma-Aldrich®
6-Hydroxy-3-coumaranon
Formel: C₈H₆O₃
Molecular Weight: 150.14 g/mol
MDL: MFCD00068174
CAS-Nummer: 6272-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR