Schweres Magnesiumkarbonat basic, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Magnesium carbonate hydroxide, Magnesium carbonate basic, heavy

M-Clarity™ Qualitätsstufe = MQ200

63032-1KG-F
SIAL63032-1KG-FEA 58.2 EUR
SIAL63032-1KG-F
Schweres Magnesiumkarbonat basic, Sigma-Aldrich®
Schweres Magnesiumkarbonat
Formel: CH₂Mg₂O₅
Molecular Weight: 142,63 g/mol
Dichte: 2,16 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011107
CAS-Nummer: 39409-82-0
EINECS: 235-192-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR