2'-Hydroxy-4',6'-dimethoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Hydroxy-4,6-dimethoxyphenyl)ethan-1-on

M-Clarity™ Qualitätsstufe = MQ100

630586-5G
SIAL630586-5GEA 61.9 EUR
SIAL630586-5G
2'-Hydroxy-4',6'-dimethoxyacetophenon, Sigma-Aldrich®
2'-Hydroxy-4',6'-dimethoxyacetophenon
Formel: C₁₀H₁₂O₄
Molecular Weight: 196.2
MDL: MFCD00017243
CAS-Nummer: 90-24-4
EINECS: 201-978-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR