2',4',6'-Trimethoxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2,4,6-Trimethoxyphenyl)ethan-1-on

M-Clarity™ Qualitätsstufe = MQ100

630594-5G
SIAL630594-5GEA 92.8 EUR
SIAL630594-5G
2',4',6'-Trimethoxyacetophenon, Sigma-Aldrich®
2',4',6'-Trimethoxyacetophenon
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,23 g/mol
MDL: MFCD00017238
CAS-Nummer: 832-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR