2-Phenyl-2',4',6'-trihydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2',4',6'-Trihydroxy-2-phenylacetophenone, 2-Phenyl-1-(2,4,6-trihydroxyphenyl)ethanone, Benzyl 2,4,6-Trihydroxyphenyl ketone

M-Clarity™ Qualitätsstufe = MQ100

630632-1G
SIAL630632-1GEA 107 EUR
SIAL630632-1G
2-Phenyl-2',4',6'-trihydroxyacetophenon, Sigma-Aldrich®
2-Phenyl-2',4',6'-trihydroxyacetophenon
Formel: C₁₄H₁₂O₄
Molecular Weight: 244,2427 g/mol
MDL: MFCD00205539
CAS-Nummer: 727-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR