2-Hexin-1-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hex-2-yn-1-ol
M-Clarity™ Qualitätsstufe = MQ100
630829-5G
SIAL630829-5GEA
73.6
EUR
SIAL630829-5G
2-Hexin-1-ol, Sigma-Aldrich®
2-Hexin-1-ol
Formel:
C₆H₁₀O Molecular Weight: 98,14 g/mol |
MDL:
MFCD00039540 CAS-Nummer: 764-60-3 EINECS: 212-128-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...