2,4-Dichlor-N-methylanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

630950-1G
SIAL630950-1GEA 88.5 EUR
SIAL630950-1G
2,4-Dichlor-N-methylanilin, Sigma-Aldrich®
2,4-Dichlor-N-methylanilin
Formel: C₇H₇Cl₂N
Molecular Weight: 176,04 g/mol
MDL: MFCD00052853
CAS-Nummer: 35113-88-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR