5-Chlorbenzimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chloro-1H-benzimidazole

M-Clarity™ Qualitätsstufe = MQ100

631248-5G
SIAL631248-5GEA 122 EUR
SIAL631248-5G
5-Chlorbenzimidazol, Sigma-Aldrich®
5-Chlorobenzimidazole
Formel: C₇H₅ClN₂
Molecular Weight: 152.581 g/mol
MDL: MFCD17015012
CAS-Nummer: 4887-82-5
EINECS: 225-503-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR