4-tert-Butylbenzylamin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

631280-25G 631280-5G
SIAL631280-25GEA 204 EUR
SIAL631280-25G SIAL631280-5G
4-tert-Butylbenzylamin, Sigma-Aldrich®
4-tert-Butylbenzylamin
Formel: C₁₁H₁₇N
MDL: MFCD00040754
CAS-Nummer: 39895-55-1
EINECS: 254-681-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR