Benzyl-4-chlorphenylketon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone

M-Clarity™ Qualitätsstufe = MQ100

631345-5G
SIAL631345-5GEA 45.7 EUR
SIAL631345-5G
Benzyl-4-chlorphenylketon, Sigma-Aldrich®
Benzyl-4-chlorphenylketon
Formel: C₁₄H₁₁ClO
Molecular Weight: 230,69 g/mol
MDL: MFCD00016342
CAS-Nummer: 1889-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR