2-Amino-3-brom-5-methylbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

631531-5G
SIAL631531-5GEA 57.2 EUR
SIAL631531-5G
2-Amino-3-brom-5-methylbenzoesäure, Sigma-Aldrich®
2-Amino-3-bromo-5-methylbenzoic acid
Formel: C₈H₈BrNO₂
Molecular Weight: 230.06
MDL: MFCD00051705
CAS-Nummer: 13091-43-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR