4-Phenyl-1-butin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

632058-5G
SIAL632058-5GEA 120 EUR
SIAL632058-5G
4-Phenyl-1-butin, Sigma-Aldrich®
4-Phenyl-1-butin
Formel: C₁₀H₁₀
Molecular Weight: 130,19 g/mol
MDL: MFCD00041667
CAS-Nummer: 16520-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR