2,2,6,6-Tetrakis(hydroxymethyl)cyclohexanol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
632082-5G
SIAL632082-5GEA
73.4
EUR
SIAL632082-5G
2,2,6,6-Tetrakis(hydroxymethyl)cyclohexanol, Sigma-Aldrich®
2,2,6,6-Tetrakis(hydroxymethyl)cyclohexanol
Formel:
C₁₀H₂₀O₅ Molecular Weight: 220,27 g/mol |
CAS-Nummer:
5416-55-7 EINECS: 226-511-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...