Methyl-4-methyl-3-nitrobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-methyl-3-nitrobenzoate

M-Clarity™ Qualitätsstufe = MQ100

632171-5G
SIAL632171-5GEA 47.3 EUR
SIAL632171-5G
Methyl-4-methyl-3-nitrobenzoat, Sigma-Aldrich®
Methyl-4-methyl-3-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,18 g/mol
MDL: MFCD00130054
CAS-Nummer: 7356-11-8
EINECS: 230-886-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR