Indium(III)oxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
632317-25G
632317-5G
SIAL632317-25GEA
362
EUR
SIAL632317-25G
SIAL632317-5G
Indium(III)oxid, Sigma-Aldrich®
Indium(III)oxid
Formel:
In₂O₃ Dichte: 7,18 g/cm³ (20 °C) |
MDL:
MFCD00011060 CAS-Nummer: 1312-43-2 EINECS: 215-193-9 Merck Index: 12,04984 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...