2-Bromnicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-3-pyridincarbonsäure, 2-Brompyridin-3-carbonsäure

M-Clarity™ Qualitätsstufe = MQ100

632465-5G
SIAL632465-5GEA 45.4 EUR
SIAL632465-5G
2-Bromnicotinsäure, Sigma-Aldrich®
2-Bromnicotinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD01646068
CAS-Nummer: 35905-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR