2-Mercapto-4-phenylthiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Phenylthiazole-2-thiol

M-Clarity™ Qualitätsstufe = MQ100

632570-5G
SIAL632570-5GEA 95 EUR
SIAL632570-5G
2-Mercapto-4-phenylthiazol, Sigma-Aldrich®
2-Mercapto-4-phenylthiazole
Formel: C₉H₇NS₂
Molecular Weight: 193,30 g/mol
MDL: MFCD00014508
CAS-Nummer: 2103-88-0
EINECS: 218-274-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR