4-Fluorphenylboronsäureneopentylglycolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-Fluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

M-Clarity™ Qualitätsstufe = MQ100

632686-1G
SIAL632686-1GEA 83.7 EUR
SIAL632686-1G
4-Fluorphenylboronsäureneopentylglycolester, Sigma-Aldrich®
4-Fluorphenylboronsäureneopentylglycolester
Formel: C₁₁H₁₄BFO₂
Molecular Weight: 208.04 g/mol
MDL: MFCD05663850
CAS-Nummer: 225916-39-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR