1-Boc-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

632732-1G 632732-5G
SIAL632732-1GEA 41.2 EUR
SIAL632732-1G SIAL632732-5G
1-Boc-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
1-Boc-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₄H₂₃N₂O₄B
MDL: MFCD05663873
CAS-Nummer: 552846-17-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR