4-Chlor-3-picolin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro-3-methylpyridine hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

632872-1G
SIAL632872-1GEA 25 EUR
SIAL632872-1G
4-Chlor-3-picolin Hydrochlorid, Sigma-Aldrich®
4-Chlor-3-picolin Hydrochlorid
Formel: C₆H₇Cl₂N
Molecular Weight: 164,04 g/mol
MDL: MFCD03092890
CAS-Nummer: 19524-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR