2-Acetyl-3-bromthiophen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(3-Brom-2-thienyl)-1-ethanon

M-Clarity™ Qualitätsstufe = MQ100

633054-25G
SIAL633054-25GEA 293 EUR
SIAL633054-25G
2-Acetyl-3-bromthiophen, Sigma-Aldrich®
2-Acetyl-3-bromthiophen
Formel: C₆H₅BrOS
Molecular Weight: 205,08 g/mol
MDL: MFCD00794287
CAS-Nummer: 42877-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR