1-Methylimidazol-5-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methylimidazole-5-carboxaldehyde, 1-Methyl-1H-imidazole-5-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

633569-1G
SIAL633569-1GEA 92.5 EUR
SIAL633569-1G
1-Methylimidazol-5-carbaldehyd, Sigma-Aldrich®
1-Methylimidazol-5-carbaldehyd
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
MDL: MFCD00030439
CAS-Nummer: 39021-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR