5,7-Dichlorisatin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

633623-1G
SIAL633623-1GEA 119 EUR
SIAL633623-1G
5,7-Dichlorisatin, Sigma-Aldrich®
5,7-Dichlorisatin
Formel: C₈H₃Cl₂NO₂
Molecular Weight: 216,02 g/mol
MDL: MFCD00043905
CAS-Nummer: 6374-92-1
EINECS: 228-928-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR