(S)-(+)-1-Boc-3-hydroxypyrrolidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

634786-5G 634786-1G
SIAL634786-5GEA 129 EUR
SIAL634786-5G SIAL634786-1G
(S)-(+)-1-Boc-3-hydroxypyrrolidin, Sigma-Aldrich®
(S)-(+)-1-Boc-3-hydroxypyrrolidin
Formel: C₉H₁₇NO₃
MDL: MFCD01317839
CAS-Nummer: 101469-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR