Magnesiumaluminat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Magnesiumaluminiumoxid, Dialuminiummagnesiumtetraoxid, Aluminiummagnesiumoxid

M-Clarity™ Qualitätsstufe = MQ100

634840-1EA
SIAL634840-1EAEA 321 EUR
SIAL634840-1EA
Magnesiumaluminat, Sigma-Aldrich®
Magnesium aluminate
Formel: Al₂MgO₄
Molecular Weight: 142,27 g/mol
MDL: MFCD00049475
CAS-Nummer: 12068-51-8
EINECS: 235-100-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR