5-Brom-2-methoxypyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

634883-1G
SIAL634883-1GEA 28.2 EUR
SIAL634883-1G
5-Brom-2-methoxypyrimidin, Sigma-Aldrich®
5-Bromo-2-methoxypyrimidine
Formel: C₅H₅BrN₂O
Molecular Weight: 189,01 g/mol
MDL: MFCD04973351
CAS-Nummer: 14001-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR