D(+)-Mannose, Millipore®
Lieferant: Merck
Synonyme:
(2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
MQ200
63580-100G
63580-25G
SIAL63580-100GEA
200
EUR
SIAL63580-100G
SIAL63580-25G
D(+)-Mannose, Millipore®
D(+)-Mannose
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol Siedepunkt: 376 Schmelzpunkt: 133 Dichte: 1.539 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064122 CAS-Nummer: 3458-28-4 EINECS: 222-392-4 Merck Index: 13,05772 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...