(2-Cyanethyl)hydrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Hydrazinylpropanenitrile

M-Clarity™ Qualitätsstufe = MQ100

636193-1G
SIAL636193-1GEA 34.4 EUR
SIAL636193-1G
(2-Cyanethyl)hydrazin, Sigma-Aldrich®
(2-Cyanoethyl)hydrazine
Formel: C₃H₇N₃
Molecular Weight: 85,11 g/mol
MDL: MFCD00043620
CAS-Nummer: 353-07-1
UN: 2927
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR