2-Amino-5-brompyrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-bromopyrazin-2-amine

M-Clarity™ Qualitätsstufe = MQ100

636320-1G
SIAL636320-1GEA 68.1 EUR
SIAL636320-1G
2-Amino-5-brompyrazin, Sigma-Aldrich®
2-Amino-5-bromopyrazine
Formel: C₄H₄BrN₃
Molecular Weight: 174.00 g/mol
MDL: MFCD00235015
CAS-Nummer: 59489-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR