5-Brom-2-hydroxy-3-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one

M-Clarity™ Qualitätsstufe = MQ100

636355-1G
SIAL636355-1GEA 67.4 EUR
SIAL636355-1G
5-Brom-2-hydroxy-3-picolin, Sigma-Aldrich®
5-Brom-2-hydroxy-3-picolin
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD03427657
CAS-Nummer: 89488-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR