5-Brom-2-hydroxy-4-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-hydroxy-4-methylpyridine

M-Clarity™ Qualitätsstufe = MQ100

636363-1G
SIAL636363-1GEA 62.7 EUR
SIAL636363-1G
5-Brom-2-hydroxy-4-picolin, Sigma-Aldrich®
5-Brom-2-hydroxy-4-picolin
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD03427664
CAS-Nummer: 164513-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR