2,4-Dichlorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

636584-5G
SIAL636584-5GEA 81.6 EUR
SIAL636584-5G
2,4-Dichlorpyridin, Sigma-Aldrich®
2,4-Dichloropyridine
Formel: C₅H₃Cl₂N
Molecular Weight: 147,99 g/mol
MDL: MFCD00955618
CAS-Nummer: 26452-80-2
EINECS: 247-717-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR