7-Benzyloxyindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-(Benzyloxy)-1H-indole

M-Clarity™ Qualitätsstufe = MQ100

636800-5G
SIAL636800-5GEA 65.7 EUR
SIAL636800-5G
7-Benzyloxyindol, Sigma-Aldrich®
7-Benzyloxyindol
Formel: C₁₅H₁₃NO
Molecular Weight: 223,27 g/mol
MDL: MFCD00037974
CAS-Nummer: 20289-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR