N-(3-Acetyl-4-hydroxyphenyl)butyramid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ac-HoPhe-OH

M-Clarity™ Qualitätsstufe = MQ100

636878-5G
SIAL636878-5GEA 64.3 EUR
SIAL636878-5G
N-(3-Acetyl-4-hydroxyphenyl)butyramid, Sigma-Aldrich®
N-(3-Acetyl-4-hydroxyphenyl)butyramid
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221,26 g/mol
MDL: MFCD00798556
CAS-Nummer: 40188-45-2
EINECS: 254-831-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR