2,6-Dichlorchinoxalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

636894-5G
SIAL636894-5GEA 118 EUR
SIAL636894-5G
2,6-Dichlorchinoxalin, Sigma-Aldrich®
2,6-Dichloroquinoxaline
Formel: C₈H₄Cl₂N₂
Molecular Weight: 199,0368 g/mol
MDL: MFCD00839695
CAS-Nummer: 18671-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR