5-Brom-2-iodpyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

637750-1G
SIAL637750-1GEA 52.6 EUR
SIAL637750-1G
5-Brom-2-iodpyrimidin, Sigma-Aldrich®
5-Bromo-2-iodopyrimidine
Formel: C₄H₂BrIN₂
MDL: MFCD01318111
CAS-Nummer: 183438-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR