2-Brom-1,3,5-triisopropylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-2,4,6-triisopropylbenzol

M-Clarity™ Qualitätsstufe = MQ300

637939-5G
SIAL637939-5GEA 34.4 EUR
SIAL637939-5G
2-Brom-1,3,5-triisopropylbenzol, Sigma-Aldrich®
2-Brom-1,3,5-triisopropylbenzol
Formel: C₁₅H₂₃Br
Molecular Weight: 283,25 g/mol
MDL: MFCD00051547
CAS-Nummer: 21524-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR