1-Brom-2-fluor-4-iodbenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

638404-5G
SIAL638404-5GEA 69.6 EUR
SIAL638404-5G
1-Brom-2-fluor-4-iodbenzol, Sigma-Aldrich®
1-Brom-2-fluor-4-iodbenzol
Formel: C₆H₃BrFI
Molecular Weight: 300,9 g/mol
MDL: MFCD00042577
CAS-Nummer: 136434-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR