4,5-Dibrom-o-xylol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

638447-5G
SIAL638447-5GEA 69 EUR
SIAL638447-5G
4,5-Dibrom-o-xylol, Sigma-Aldrich®
4,5-Dibrom-o-xylol
Formel: C₈H₈Br₂
MDL: MFCD00082743
CAS-Nummer: 24932-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR