2-(Tributylstannyl)oxazol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
638617-1G
638617-5G
SIAL638617-1GEA
230
EUR
SIAL638617-1G
SIAL638617-5G
2-(Tributylstannyl)oxazol, Sigma-Aldrich®
2-(Tributylstannyl)oxazol
Formel:
C₁₅H₂₉NOSn |
MDL:
MFCD06798076 CAS-Nummer: 145214-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...