2-Chlor-3,4-dimethoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

638730-5G
SIAL638730-5GEA 238 EUR
SIAL638730-5G
2-Chlor-3,4-dimethoxybenzoesäure, Sigma-Aldrich®
2-Chloro-3,4-dimethoxybenzoic acid
Formel: C₉H₉ClO₄
Molecular Weight: 216,62 g/mol
MDL: MFCD00084944
CAS-Nummer: 52009-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR