Mercury disposal kit, Mercurisorb™
Lieferant: Merck
63879-1EA
SIAL63879-1EAEA 218 EUR
SIAL63879-1EA
Mercury disposal kit, Mercurisorb™
Absorptionsmaterial
Rapid absorber for safe disposal of mercury.
Lieferumfang: The kit includes disposable gloves, minishovel and brush, disposable pipettes in a compact plastic case.