4-Brom-2,6-dimethylanisol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
638803-5G
SIAL638803-5GEA
82.6
EUR
SIAL638803-5G
4-Brom-2,6-dimethylanisol, Sigma-Aldrich®
4-Brom-2,6-dimethylanisol
Formel:
C₉H₁₁BrO Molecular Weight: 215,09 g/mol Siedepunkt: 117…120 °C (13 mmHg) |
MDL:
MFCD00143259 CAS-Nummer: 14804-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...