3,5-Dimethyl-4-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methoxy-3,5-dimethylbenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

638811-5G
SIAL638811-5GEA 147 EUR
SIAL638811-5G
3,5-Dimethyl-4-methoxybenzonitril, Sigma-Aldrich®
3,5-Dimethyl-4-methoxybenzonitril
Formel: C₁₀H₁₁NO
Molecular Weight: 161,2004 g/mol
MDL: MFCD03428544
CAS-Nummer: 152775-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR