4-Brom-5-fluor-2-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

638994-5G
SIAL638994-5GEA 123 EUR
SIAL638994-5G
4-Brom-5-fluor-2-nitrotoluol, Sigma-Aldrich®
4-Brom-5-fluor-2-nitrotoluol
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,02 g/mol
MDL: MFCD04039299
CAS-Nummer: 224185-19-7
EINECS: 262-536-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR